Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCN8, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89129002 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-002 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-002 Supplier Abnova Corporation Supplier No. PAB27490

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit polyclonal antibody raised against recombinant LCN8.

Members of the lipocalin family, such as LCN8, have a common structure consisting of an 8-stranded antiparallel beta-barrel that forms a cup-shaped ligand-binding pocket or calyx. Lipocalins generally bind small hydrophobic ligands and transport them to specific cells (Suzuki et al., 2004 [PubMed 15363845]).[supplied by OMIM

Sequence: IGGFWREVGVASDQSLVLTAPKRVEGLFLTLSGSNLTVKVAYNSSGSCEIEKIVGSEIDSTG

Specifications

Antigen LCN8
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description lipocalin 8
Formulation In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide)
Gene LCN8
Gene Alias EP17, LCN5
Gene Symbols LCN8
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human LCN8.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138307
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.