Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-LDHB, Polyclonal,

Catalog No. ABC927MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC927MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC927MI Supplier MilliporeSigma™ Supplier No. ABC927
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects LDHB in Human samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

L-lactate dehydrogenase B chain (EC 1.1.1.27; UniProt P07195; also known as Epididymis secretory protein Li 281, Lactate dehydrogenase H chain, LDH-B, LDH-H, LDH heart subunit, Renal carcinoma antigen NY-REN-46) is encoded by the LDHB (also known as LDHBD, TRG-5, HEL-S-281) gene (Gene ID 3945) in human. Originally described by Otto Warburg and now commonly known as the Warburg effect refers to the phenomenon that many tumor cells use aerobic glycolysis instead of mitochondrial respiration for energy production to support cell growth and proliferation. Enzymes involved in the complex network of metabolic pathways are therefore being studied as potential targets for cancer treatment. Lactate dehydrogenase (LDH) is a tetrameric enzyme composed of two different subunits, LDHA and LDHB, which can assemble into five different combinations (LDH1 is comprised of four LDHB subunits; LDH2 contains three LDHB and one LDHA subunit; LDH3 contains two LDHB and two LDHA subunits; LDH4 contains one LDHB and three LDHA subunits; LDH5 contains four LDHA subunits). LDHB is ubiquitously expressed and the predominant isoform in heart muscle, while LDHA is the predominant isoform in skeletal muscle and other highly glycolytic tissues. LDH catalyzes the reversible conversion of pyruvate into lactate. An upregulation of LDH therefore reduces the oxygen dependency and ensures an efficient anaerobic/glycolytic metabolism in tumor cells. Serum LDHB levels, for example, are reported to elevate in lung cancer patients and progressively increase with the clinical stage of non-small cell lung cancer (NSCLC).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDHB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P07195
Gene Symbols LDHB
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human LDHB sequence from the C-terminal half.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_002291
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.