Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEFTY2, Rabbit, Clone: 7C5G1H6H10, Abnova™

Catalog No. 89110774 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-774 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-774 Supplier Abnova Corporation Supplier No. MAB2032
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against recombinant LEFTY2.

This gene encodes a member of the TGF-beta family of proteins. The encoded protein is secreted and plays a role in left-right asymmetry determination of organ systems during development. The protein may also play a role in endometrial bleeding. Mutations in this gene have been associated with left-right axis malformations, particularly in the heart and lungs. Some types of infertility have been associated with dysregulated expression of this gene in the endometrium. Alternative processing of this protein can yield three different products. This gene is closely linked to both a related family member and a related pseudogene. [provided by RefSeq

Specifications

Antigen LEFTY2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 7C5G1H6H10
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against recombinant LEFTY2.
Dilution ELISA (1:5000) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene LEFTY2
Gene Alias EBAF/LEFTA/LEFTYA/MGC46222/TGFB4
Gene Symbols LEFTY2
Host Species Rabbit
Immunogen Recombinant His fusion protein corresponding to human LEFTY2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7044
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.