Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEPR Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89116814 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-814 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-814 Supplier Abnova Corporation Supplier No. PAB13709
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Lepr.

Sequence: KEIQWKTHEVFDAKSKSA

Specifications

Antigen LEPR
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Lepr.
Dilution Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene Lepr
Gene Alias Fa
Gene Symbols Lepr
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 577-594 of rat Lepr.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24536
Test Specificity Recognizes leptin receptor. Detects bands of 100 KDa and 125 KDa, representing the short and long forms of leptin receptor. Also detects two unrelated bands of 31 KDa and 35 KDa.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.