Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-LILRB2, Polyclonal,

Catalog No. ABN1023MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1023MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1023MI Supplier MilliporeSigma™ Supplier No. ABN1023
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects LILRB2 in Human samples, and it is validated for Immunohistochemistry, Western Blotting

Leukocyte immunoglobulin-like receptor subfamily B member 2 (UniProt Q8N423; also known as CD85 antigen-like family member D, CD85d, Ig-like transcript 4, ILT-4, Leukocyte immunoglobulin-like receptor 2, LIR-2, MIR-10, Monocyte/macrophage immunoglobulin-like receptor 10) is encoded by the LILRB2 (also known as CD85D, ILT4, LIR2, MIR10) gene (Entrez Gene ID 10288) in human. CD85d belongs to the B class of leukocyte immunoglobulin-like (LIR) receptors family, members of which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and two to four cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). Cd85d is expressed on immune cells and negatively regulates immune response by engaging class I MHC molecules (HLA-A, HLA-B, HLA-C) on antigen-presenting cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LILRB2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q8N423
Gene Symbols LILRB2; ILT4; LIR2; MIR10
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the extracellular domain of human LILRB2.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_005865
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.