Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lime1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89116734 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-734 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-734 Supplier Abnova Corporation Supplier No. PAB13550

Rabbit polyclonal antibody raised against partial recombinant Lime1.

Specifications

Antigen Lime1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Lime1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene Lime1
Gene Alias 2810038K19Rik/LIME
Gene Symbols Lime1
Host Species Rabbit
Immunogen Recombinant His fusion protein corresponding to amino acids 86-202 of mouse Lime1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72699
Test Specificity This antibody reacts with the cytoplasmic domain of mouse Lime1 (Lck-interacting molecule), a transmembrane adaptor protein involved in regulation of T cell activation by coreceptors.
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.