Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIPE Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89115602 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-602 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-602 Supplier Abnova Corporation Supplier No. PAB12105
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Lipe.

Specifications

Antigen LIPE
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Lipe.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (1mg/mL BSA, 0.05% sodium azide)
Gene Lipe
Gene Alias HSL
Gene Symbols Lipe
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 699-908 of rat Lipe.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25330
Test Specificity This antibody is specific to hormone-sensitive lipase protein.
Target Species Human, Rat
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.