Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMP-1, Rabbit, Clone: D24-G, Abnova™

Catalog No. 89113773 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-773 Supplier Abnova Corporation Supplier No. MAB9772
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against synthetic peptide of LMP-1.

Specifications

Antigen LMP-1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone D24-G
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of LMP-1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:200) The optimal working dilution should be determined by the end user.
Formulation In 20mM Tris-HCl, pH 8.0 (20mg/mL BSA, 0.05% sodium azide)
Gene LMP-1
Gene Accession No. P03230
Gene Symbols LMP-1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to LMP-1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3783750
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.