Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMP-1, Rabbit, Clone: S20-D, Abnova™
Rabbit monoclonal antibody raised against synthetic peptide of LMP-1.
Supplier: Abnova Corporation MAB9773
Specifications
LMP-1 | |
Monoclonal | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:200) The optimal working dilution should be determined by the end user. | |
LMP-1 | |
LMP-1 | |
A synthetic peptide corresponding to C-terminus of Epstein-Barr virus LMP-1. | |
RUO | |
3783750 | |
Store at 4°C. |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
S20-D | |
Rabbit monoclonal antibody raised against synthetic peptide of LMP-1. | |
In 20mM Tris-HCl, pH 8.0 (20mg/mL BSA, 0.05% sodium azide) | |
P03230 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction