Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lox Rabbit anti-Human, Mouse, Pig, Primates, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89137486 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-486 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-486 Supplier Abnova Corporation Supplier No. PAB12158
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Lox.

Specifications

Antigen Lox
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Lox.
Dilution Immunocytochemistry (1:50) Immunofluorescence (1:50) Immunohistochemistry (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:250) The optimal working dilution should be determined by the end user.
Formulation In PBS
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 78-115 of mouse Lox.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes the pro-enzyme (50 KDa) and pro-peptide. The pro-peptide runs at ∼18-20 KDa when not glycosylated and ∼35 KDa on SDS-PAGE when N-glycosylated.
Target Species Human, Mouse, Pig, Primate, Rat
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.