Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LPO Rabbit anti-Bovine, Human, Primates, Polyclonal Antibody, Abnova™

Catalog No. 89115773 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-773 Supplier Abnova Corporation Supplier No. PAB12397
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of LPO.

This gene encodes an oxidoreductase secreted from salivary, mammary, and other mucosal glands that functions as a natural antibacterial agent. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen LPO
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of LPO.
Dilution Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (30% glycerol, 0.09% sodium azide)
Gene LPO
Gene Alias MGC129990/MGC129991/SPO
Gene Symbols LPO
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 300-400 of human LPO.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4025
Target Species Bovine, Human, Primate
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.