Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRBA Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89123090 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-090 Supplier Abnova Corporation Supplier No. PAB21593
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant LRBA.

Patients with Chediak-Higashi syndrome (CHS1; MIM 214500) suffer from a systemic immunodeficiency involving defects in polarized trafficking of vesicles in a number of immune system cell types. In mouse, this syndrome is reproduced in strains with a mutation in the 'beige' gene that results in proteins lacking the BEACH (beige and CHS1) domain and C-terminal WD repeats. LRBA contains key features of both beige/CHS1 and A kinase anchor proteins (AKAPs; see MIM 602449).[supplied by OMIM

Sequence: GGWRVWVDTLSITHSKVTFEIHKENLANIFREQQGKVDEEIGLCSSTSVQAASGIRRDINVSVGSQQPDTKDSPVCPHFTTNGNENSSIEKTSSLESASNIELQTTNTSYEEMKAEQENQELPDEGTLEETL

Specifications

Antigen LRBA
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant LRBA.
Dilution Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene LRBA
Gene Alias BGL/CDC4L/DKFZp686A09128/DKFZp686K03100/DKFZp686P2258/FLJ16600/FLJ25686/LAB300/LBA/MGC72098
Gene Symbols LRBA
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human LRBA.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 987
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.