Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRP6 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89116032 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-032 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-032 Supplier Abnova Corporation Supplier No. PAB12700
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of LRP6.

The LRP6 gene encodes a member of the low density lipoprotein receptor (LDLR) gene family, which consists of cell surface proteins involved in receptor-mediated endocytosis of specific ligands. LDLR proteins are comprised of the same basic structural motifs: an extracellular domain that contains LDLR binding repeats and EGF repeats with associated spacer domains containing the YWTD motif; a single transmembrane-spanning domain; and a C-terminal cytoplasmic domain that generally contains at least 1 copy of the NPXY motif (Brown et al., 1998 [PubMed 9704021]). LRP6 and LRP5 (MIM 603506) function as coreceptors for WNT (MIM 164820) ligands and thus play a role in WNT signaling, which is important in a wide variety of biologic processes during pre- and postnatal life in invertebrates and vertebrates (Kokubu et al., 2004 [PubMed 15469977]).[supplied by OMIM

Specifications

Antigen LRP6
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of LRP6.
Dilution Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In TBS, pH 7.2 (BSA, 10% Proclin300)
Gene LRP6
Gene Alias ADCAD2/FLJ90062/FLJ90421
Gene Symbols LRP6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids (KGTYF) of human LRP6.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4040
Test Specificity This antibody recognizes ∼180 KDa of human LRP6.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.