Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Luteinizing Hormone-Releasing Hormone, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89138220 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-220 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-220 Supplier Abnova Corporation Supplier No. PAB16667

Rabbit polyclonal antobody raised against synthetic peptide of Luteinizing Hormone-Releasing Hormone.

Specifications

Antigen Luteinizing Hormone-Releasing Hormone
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antobody raised against synthetic peptide of Luteinizing Hormone-Releasing Hormone.
Dilution Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulation In serum
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with BSA) corresponding to Luteinizing Hormone-Releasing Hormone.
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.