Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LY6K Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114876 Shop All Abnova Corporation Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-876 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. 89-114-876 missing translation for 'mfr' Abnova Corporation missing translation for 'supplierNo' PAB1086

Rabbit polyclonal antibody raised against synthetic peptide of LY6K.

Sequence: RQRDPEDSQRTDEGDNRVW

Specifications

Antigen LY6K
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description lymphocyte antigen 6 complex, locus K
Formulation In PBS (0.02% sodium azide)
Gene LY6K
Gene Alias FLJ35226, HSJ001348
Gene Symbols LY6K
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 82-100 of human LY6K.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54742
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.