Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-LY75, Clone: R18-D, Abnova™

Catalog No. 89110548 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-548 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-548 Supplier Abnova Corporation Supplier No. MAB1651

Specifications

Antigen LY75
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone R18-D
Conjugate Unconjugated
Formulation In 20mM Tris-HCl, pH 8.0 (20mg/mL BSA, 0.05% sodium azide)
Gene Symbols LY75
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human LY75.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 4065
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.