Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYNX1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89123098 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-098 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-098 Supplier Abnova Corporation Supplier No. PAB21601

Rabbit polyclonal antibody raised against recombinant LYNX1.

This gene encodes a member of the Ly-6/neurotoxin gene family, a group of lymphocyte antigens that attach to the cell surface by a glycosylphosphatidylinositol anchor and have a unique structure showing conserved 8-10 cysteine residues with a characteristic spacing pattern. Functional analysis indicates that this protein is not a ligand or neurotransmitter but has the capacity to enhance nicotinic acetylcholine receptor function in the presence of acetylcholine. This gene may also play a role in the pathogenesis of psoriasis vulgaris. Alternatively spliced variants encoding different isoforms have been identified. [provided by RefSeq

Sequence: IWCHQCTGFGGCSHGSRCLRDSTHCVTTATRVLSNTEDLPLVTKMCHIGCPDIPSLGLGPYVSIACCQTSLCNHD

Specifications

Antigen LYNX1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Ly6/neurotoxin 1
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene LYNX1
Gene Alias MGC40364, SLURP2
Gene Symbols LYNX1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human LYNX1.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66004
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.