Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYVE1, Rabbit, DyLight 549, Polyclonal Antibody, Abnova™

Catalog No. 89117398 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-398 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-398 Supplier Abnova Corporation Supplier No. PAB14998
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Lyve1.

Specifications

Antigen LYVE1
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate DyLight 549
Description Rabbit polyclonal antibody raised against synthetic peptide of Lyve1.
Dilution Flow Cytometry (1:25) The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium borate
Gene Lyve1
Gene Alias 1200012G08Rik/Crsbp-1/Lyve-1/Xlkd1
Gene Symbols Lyve1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 250-318 of mouse Lyve1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114332
Test Specificity This antibody cross-reacts with human and mouse. No other species have been tested. Rabbit Polyclonal to LYVE1 Western blot where a band at ∼45 KDa is seen.
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.