Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

M6prbp1 (Plin3), Rabbit, HRP, Polyclonal Antibody, Abnova™

Catalog No. 89117470 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-470 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-470 Supplier Abnova Corporation Supplier No. PAB15143
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Plin3.

Specifications

Antigen M6prbp1 (Plin3)
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Description Rabbit polyclonal antibody raised against synthetic peptide of Plin3.
Dilution Western Blot (1:1000-1:5000) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene M6prbp1
Gene Alias 1300012C15Rik/Tip47
Gene Symbols M6prbp1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1-100 of mouse Plin3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66905
Test Specificity TIP47 This antibody is useful for Western blot analysis where a band is seen ∼47 KDa (isoform B, containing the first 184 residues that isoform A is lacking).
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.