Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGEB17, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant MAGEB17.
Supplier: Abnova Corporation PAB27472
Description
Sequence: LYLKEMDPSRQSYVLVGKLDFPNQGSLSDGGGFPLSGLLMVLLSTIFMHGNRATEEEMWECLNALGMYKGRKHFIYGEPQELVTKDLVREGYLEYQQVPSSDPPRYEFLWGPRARAETSKMKVSpecifications
| MAGEB17 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant MAGEB17. | |
| In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide) | |
| MAGEB17 | |
| Recombinant protein corresponding to amino acids of human MAGEB17. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Immunohistochemistry (Formalin/Paraffin) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| MAGEB17 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 645864 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction