Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP1LC3A Rabbit anti-Bovine, Human, Mouse, Rat, Zebra fish, Polyclonal Antibody, Abnova™

Catalog No. 89117631 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-631 Supplier Abnova Corporation Supplier No. PAB15521
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against MAP1LC3A.

MAP1A and MAP1B are microtubule-associated proteins which mediate the physical interactions between microtubules and components of the cytoskeleton. MAP1A and MAP1B each consist of a heavy chain subunit and multiple light chain subunits. The protein encoded by this gene is one of the light chain subunits and can associate with either MAP1A or MAP1B. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen MAP1LC3A
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against MAP1LC3A.
Dilution Flow Cytometry (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Western Blot (1:2500) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.1% BSA, 0.05% sodium azide)
Gene MAP1LC3A
Gene Alias LC3/LC3A/MAP1ALC3/MAP1BLC3
Gene Symbols MAP1LC3A
Host Species Rabbit
Immunogen N-terminus of human MAP1LC3A.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84557
Test Specificity Immunogen has 100% homology to rat and 97% homology to bovine. This antibody is useful for Western blot, where a band is seen ∼14 KDa.
Target Species Bovine, Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.