Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2K1/MAP2K2 (phospho S218/S222), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89115969 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-969 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-969 Supplier Abnova Corporation Supplier No. PAB12635
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAP2K1/MAP2K2.

Sequence: SMANS

Specifications

Antigen MAP2K1/MAP2K2
Applications Dot Blot, ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAP2K1/MAP2K2.
Dilution Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In TBS, pH 7.2 (BSA, 10% Proclin300)
Gene Alias MAPKK1/MEK1/MKK1/PRKMK1
Gene Symbols MAP2K1
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S218/S222 of human MAP2K1/MAP2K2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5604/5605
Test Specificity This antibody recognizes ∼44/45 KDa of human MAP2K1/MAP2K2 at the phosphorylation site of Ser 218/222 or MEK2(pS222/226). It does not cross react to non-phospho MEK1/2.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C or lower.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.