Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Map2k4, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89133965 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-133-965 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-133-965 Supplier Abnova Corporation Supplier No. PAB8830

Rabbit polyclonal antibody raised against synthetic peptide of Map2k4.

Specifications

Antigen Map2k4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description mitogen-activated protein kinase kinase 4
Formulation In PBS (30% glycerol, 0.5% BSA, 0.01% thimerosal)
Gene Map2k4
Gene Alias JNKK1, MEK4, MKK4, PRKMK4, Sek1, Serk1
Gene Symbols Map2k4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues surrounding amino acids 347 of mouse Map2k4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26398
Test Specificity This antibody recognizes unphosphorylated and phosphorylated SEK1.
Target Species Canine, Human, Mouse
Content And Storage Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.