Learn More
MAP2K4 (phospho S80), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAP2K4.
Supplier: Abnova Corporation PAB8908
Description
This gene encodes a dual specificity protein kinase that belongs to the Ser/Thr protein kinase family. This kinase is a direct activator of MAP kinases in response to various environmental stresses or mitogenic stimuli. It has been shown to activate MAPK8/JNK1, MAPK9/JNK2, and MAPK14/p38, but not MAPK1/ERK2 or MAPK3/ERK3. This kinase is phosphorylated, and thus activated by MAP3K1/MEKK. The knockout studies in mice suggested the roles of this kinase in mediating survival signal in T cell development, as well as in the organogenesis of liver. [provided by RefSeq
Specifications
MAP2K4 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAP2K4. | |
In PBS (50% glycerol, 1.0% BSA, 0.02% thimerosal) | |
JNKK/JNKK1/MAPKK4/MEK4/MKK4/PRKMK4/SEK1/SERK1 | |
Rabbit | |
100 μg | |
Primary | |
This antibody recognizes phosphorylated SEK1/MKK4/JKK1. | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
Western Blot (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
MAP2K4 | |
MAP2K4 | |
Synthetic phosphopeptide corresponding to residues surrounding S80 of human MAP2K4. | |
RUO | |
6416 | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only