Learn More
MAP3K11 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MAP3K11.
Supplier: Abnova Corporation PAB8841
Description
The protein encoded by this gene is a member of the serine/threonine kinase family. This kinase contains a SH3 domain and a leucine zipper-basic motif. This kinase preferentially activates MAPK8/JNK kinase, and functions as a positive regulator of JNK signaling pathway. This kinase can directly phosphorylate, and activates IkappaB kinase alpha and beta, and is found to be involved in the transcription activity of NF-kappaB mediated by Rho family GTPases and CDC42. [provided by RefSeq
Specifications
MAP3K11 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of MAP3K11. | |
In PBS (50% glycerol, 1.0% BSA, 0.02% thimerosal) | |
MGC17114/MLK-3/MLK3/PTK1/SPRK | |
Rabbit | |
100 μg | |
Primary | |
This antibody recognizes 92 KDa MLK3. It does not cross-react with MLK1, MLK2 and other mixed lineage kinases. | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
Western Blot (0.5-4 ug/mL) The optimal working dilution should be determined by the end user. | |
MAP3K11 | |
MAP3K11 | |
A synthetic peptide corresponding to C-terminus of human MAP3K11. | |
RUO | |
4296 | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only