Learn More
MAP4K4 (phospho S801), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAP4K4.
Supplier: Abnova Corporation PAB8118
Description
The protein encoded by this gene is a member of the serine/threonine protein kinase family. This kinase has been shown to specifically activate MAPK8/JNK. The activation of MAPK8 by this kinase is found to be inhibited by the dominant-negative mutants of MAP3K7/TAK1, MAP2K4/MKK4, and MAP2K7/MKK7, which suggests that this kinase may function through the MAP3K7-MAP2K4-MAP2K7 kinase cascade, and mediate the TNF-alpha signaling pathway. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Specifications
MAP4K4 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAP4K4. | |
In PBS (0.09% sodium azide) | |
FLH21957/FLJ10410/FLJ20373/FLJ90111/HGK/KIAA0687/NIK | |
Rabbit | |
Peptide affinity purification | |
RUO | |
9448 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Dot Blot, ELISA | |
Unconjugated | |
ELISA (1:1000) Dot Blot (1:500) The optimal working dilution should be determined by the end user. | |
MAP4K4 | |
MAP4K4 | |
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S801 of human MAP4K4. | |
400 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only