Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK1, Rabbit, Clone: B20-U, Abnova™

Catalog No. 89110535 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-535 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-535 Supplier Abnova Corporation Supplier No. MAB1636
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against synthetic peptide of MAPK1.

The protein encoded by this gene is a member of the MAP kinase family. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. Upon activation, this kinase translocates to the nucleus of the stimulated cells, where it phosphorylates nuclear targets. Two alternatively spliced transcript variants encoding the same protein, but differing in the UTRs, have been reported for this gene. [provided by RefSeq

Specifications

Antigen MAPK1
Applications ELISA, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone B20-U
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of MAPK1.
Dilution Western Blot (1:5000) ELISA (1:20000-1:100000) The optimal working dilution should be determined by the end user.
Formulation In 20mM Tris-HCl, pH 8.0 (10mg/mL BSA, 0.05% sodium azide)
Gene MAPK1
Gene Alias ERK/ERK2/ERT1/MAPK2/P42MAPK/PRKM1/PRKM2/p38/p40/p41/p41mapk
Gene Symbols MAPK1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human MAPK1.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5594
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.