Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK1/MAPK3, Rabbit, Clone: G15-B, Abnova™

Catalog No. 89110536 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-536 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-536 Supplier Abnova Corporation Supplier No. MAB1637
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against synthetic phosphopeptide of MAPK1/MAPK3.

Specifications

Antigen MAPK1/MAPK3
Applications ELISA, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone G15-B
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic phosphopeptide of MAPK1/MAPK3.
Dilution ELISA (1:20000-1:100000) Immunofluorescence Immunoprecipitation Western Blot (1:5000) The optimal working dilution should be determined by the end user.
Formulation In 20mM Tris-HCl, pH 8.0 (10mg/mL BSA, 0.05% sodium azide)
Gene Alias ERK/ERK2/ERT1/MAPK2/P42MAPK/PRKM1/PRKM2/p38/p40/p41/p41mapk
Gene Symbols MAPK1
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding T202/T204 of human MAPK1/MAPK3.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5594/5595
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.