Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAPK1/MAPK3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MAPK1/MAPK3.
Supplier: Abnova Corporation PAB26775
Description
Sequence: T-E-Yp-V-ASpecifications
MAPK1/MAPK3 | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
ERK/ERK2/ERT1/MAPK2/P42MAPK/PRKM1/PRKM2/p38/p40/p41/p41mapk | |
Rabbit | |
Affinity chromatography | |
RUO | |
5594/5595 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of MAPK1/MAPK3. | |
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
MAPK1 | |
A synthetic peptide corresponding to residues surrounding Y204 of human MAPK3/MAPK1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction