Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Mapk1, Polyclonal, Abnova™

Catalog No. 89119527 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-527 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-527 Supplier Abnova Corporation Supplier No. PAB17866

Specifications

Antigen Mapk1
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS pH 7.2 (50% glycerol, 1% BSA, 0.02% thimerosal)
Gene Symbols Mapk1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to rat Mapk1.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 116590
Target Species Chicken, Human, Mouse, Rat, Sheep
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.