Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAPK1 Rabbit anti-Dog, Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MAPK1.
Supplier: Abnova Corporation PAB12764
Specifications
MAPK1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of MAPK1. | |
In TBS, pH 7.2 (BSA, 10% Proclin300) | |
A synthetic peptide corresponding to residues surrounding T202 and Y204 of human, mouse, rat and dog MAPK1. | |
RUO | |
This antibody recognizes MAPK1/MAPK3 without the dual phosphorylation sites Thr202 and Tyr 204. | |
Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
Rabbit | |
100 μg | |
Primary | |
Canine, Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction