Learn More
MAPK14 (phospho T180/Y182), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAPK14.
Supplier: Abnova Corporation PAB8911
Description
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This kinase is activated by various environmental stresses and proinflammatory cytokines. The activation requires its phosphorylation by MAP kinase kinases (MKKs), or its autophosphorylation triggered by the interaction of MAP3K7IP1/TAB1 protein with this kinase. The substrates of this kinase include transcription regulator ATF2, MEF2C, and MAX, cell cycle regulator CDC25B, and tumor suppressor p53, which suggest the roles of this kinase in stress related transcription and cell cycle regulation, as well as in genotoxic stress response. Four alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq
Specifications
MAPK14 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAPK14. | |
In PBS (50% glycerol, 1.0% BSA, 0.02% thimerosal) | |
CSBP1/CSBP2/CSPB1/EXIP/Mxi2/PRKM14/PRKM15/RK/SAPK2A/p38/p38ALPHA | |
Rabbit | |
100 μg | |
Primary | |
This antibody recognizes 43 KDa phosphorylated p38 (Thr180/Tyr182) MAP kinase. | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Western Blot (0.5-4 ug/mL) Immunoprecipitation (10-20 mg/mL) Immunohistochemistry (10-20 ug/mL) The optimal working dilution should be determined by the end user. | |
MAPK14 | |
MAPK14 | |
Synthetic phosphopeptide corresponding to residues surrounding T180/Y182 of human MAPK14. | |
RUO | |
1432 | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only