Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mapk9, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115477 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-477 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-477 Supplier Abnova Corporation Supplier No. PAB11964

Rabbit polyclonal antibody raised against synthetic peptide of Mapk9.

Sequence: MSDSKSDGQFYSVQVA

Specifications

Antigen Mapk9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Mapk9.
Dilution Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In antiserum
Gene Mapk9
Gene Alias SAPK
Gene Symbols Mapk9
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1-16 of rat Mapk9.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50658
Test Specificity This antibody detects a 55 KDa protein on rat brain tissue extract SDS-PAGE immunoblots, corresponding to the molecular weight of SAPKalpha. A 46 KDa protein is also seen in human tissue extracts. Due to alternative splicing of the 46 and 55 KDa transcripts, numerous antigen specific bands are seen with this molecular weight range.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.