Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPKAP1 Rabbit anti-Birds, Bovine, Chicken, Dog, Human, Mouse, Rat, Zebra fish, Polyclonal Antibody, Abnova™

Catalog No. 89114111 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-111 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-111 Supplier Abnova Corporation Supplier No. PAB0290
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant MAPKAP1.

This gene encodes a protein that is highly similar to the yeast SIN1 protein, a stress-activated protein kinase. Alternatively spliced transcript variants encoding distinct isoforms have been described. Alternate polyadenylation sites as well as alternate 3' UTRs have been identified for transcripts of this gene. [provided by RefSeq]

Sequence: NTAQRLERLRKERQNQIK

Specifications

Antigen MAPKAP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant MAPKAP1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% gelatin, 0.05% sodium azide)
Gene MAPKAP1
Gene Alias JC310/MGC2745/MIP1/SIN1/SIN1b/SIN1g
Gene Symbols MAPKAP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 85-102 of human MAPKAP1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79109
Target Species Avian, Bovine, Canine, Chicken, Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.