Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-MAPKAPK2, Polyclonal, Abnova™

Catalog No. 89121137 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-137 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-137 Supplier Abnova Corporation Supplier No. PAB1961

Specifications

Antigen MAPKAPK2
Applications Dot Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS (0.09% sodium azide)
Gene Symbols MAPKAPK2
Host Species Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S272 of human MAPKAPK2.
Purification Method Protein G purification
Quantity 400 μL
Primary or Secondary Primary
Gene ID (Entrez) 9261
Target Species Human
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.