Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPT Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89126489 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-126-489 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-126-489 Supplier Abnova Corporation Supplier No. PAB24818
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of MAPT.

This gene encodes the microtubule-associated protein tau (MAPT) whose transcript undergoes complex, regulated alternative splicing, giving rise to several mRNA species. MAPT transcripts are differentially expressed in the nervous system, depending on stage of neuronal maturation and neuron type. MAPT gene mutations have been associated with several neurodegenerative disorders such as Alzheimer's disease, Pick's disease, frontotemporal dementia, cortico-basal degeneration and progressive supranuclear palsy. [provided by RefSeq]

Specifications

Antigen MAPT
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of MAPT.
Dilution Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In 1X PBS, pH 7.2 (0.05% sodium azide)
Gene MAPT
Gene Alias DDPAC/FLJ31424/FTDP-17/MAPTL/MGC138549/MSTD/MTBT1/MTBT2/PPND/TAU
Gene Symbols MAPT
Host Species Rabbit
Immunogen A synthetic peptide corresponding to MAPT.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4137
Test Specificity MAPT polyclonal antibody detects endogenous levels of MAPT protein.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.