Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAPT (phospho S416), Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of Mapt.
Supplier: Abnova Corporation PAB9655
Specifications
| MAPT | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of Mapt. | |
| In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA) | |
| MGC156663/Mtapt/RNPTAU/pTau | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 29477 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Mapt | |
| Mapt | |
| Synthetic phosphopeptide corresponding to residues surrounding S416 of rat Mapt. | |
| 100 μL | |
| Primary | |
| Bovine, Canine, Human, Mouse, Primate, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction