Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MARCH8, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MARCH8.
Supplier: Abnova Corporation PAB16567
Specifications
| MARCH8. | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MARCH8. | |
| In PBS (0.02% sodium azide) | |
| MARCH-VIII/MIR/RNF178/c-MIR | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| MARCH8 | |
| MARCH8 | |
| A synthetic peptide corresponding to C-terminus 17 amino acids of human MARCH8. | |
| RUO | |
| 220972 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction