Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MARCKS Rabbit anti-Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against full-length recombinant MARCKS.
Supplier: Abnova Corporation PAB12564
Specifications
MARCKS | |
Polyclonal | |
Rabbit polyclonal antibody raised against full length recombinant MARCKS. | |
In antiserum | |
Recombinant protein corresponding to full length MARCKS. | |
RUO | |
The MARCKS protein runs at about 80KDa on SDS-PAGE gels, although the real molecular weight is much lower, at about 31.5KDa. The reason for this is probably because the protein is extremely rich in acidic residues. Such proteins appear to bind less SDS than the average protein, and so migrate in PAGE more slowly than the normal protein. | |
Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
Immunocytochemistry (1:5000) Immunofluorescence (1:500-1:1000) Western Blot (1:20000) The optimal working dilution should be determined by the end user. | |
Rabbit | |
100 μL | |
Primary | |
Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction