Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARCKS Rabbit anti-Human, Mammals, Polyclonal Antibody, Abnova™

Catalog No. 89116967 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-967 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-967 Supplier Abnova Corporation Supplier No. PAB14167
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant MARCKS.

The protein encoded by this gene is a substrate for protein kinase C. It is localized to the plasma membrane and is an actin filament crosslinking protein. Phosphorylation by protein kinase C or binding to calcium-calmodulin inhibits its association with actin and with the plasma membrane, leading to its presence in the cytoplasm. The protein is thought to be involved in cell motility, phagocytosis, membrane trafficking and mitogenesis. [provided by RefSeq

Specifications

Antigen MARCKS
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant MARCKS.
Dilution Western Blot (1:10000) Immunofluorescence (1:1000) The optimal working dilution should be determined by the end user.
Formulation In serum (5mM sodium azide)
Gene MARCKS
Gene Alias 80K-L/FLJ14368/FLJ90045/MACS/PKCSL/PRKCSL
Gene Symbols MARCKS
Host Species Rabbit
Immunogen Recombinant protein corresponding to human MARCKS.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4082
Target Species Human, Mammalia
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.