Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARK2 (phospho T595), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114795 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-795 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-795 Supplier Abnova Corporation Supplier No. PAB10332
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of MARK2.

This gene encodes a member of the Par-1 family of serine/threonine protein kinases. The protein is an important regulator of cell polarity in epithelial and neuronal cells, and also controls the stability of microtubules through phosphorylation and inactivation of several microtubule-associating proteins. The protein localizes to cell membranes. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen MARK2
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of MARK2.
Dilution ELISA (1:1000-1:3000) Western Blot (1:250-1:1000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene MARK2
Gene Alias EMK1/MGC99619/PAR-1/Par1b
Gene Symbols MARK2
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding T595 of human MARK2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2011
Test Specificity Reactivity occurs against human MARK2 pT595 protein and This antibody is specific to the phosphorylated form of the protein. Reactivity with non-phosphorylated human MARK2 is minimal by ELISA. Expect at least partial reactivity with other isoforms of MARK. The immunogen sequence is present in all forms identified to date and localizes to T595 on MARK2, T587 on MARK3, T591 on MARK1 and T568 on MARK4.
Target Species Bovine, Canine, Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.