Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAT2A Rabbit anti-Bovine, Chicken, Human, Mouse, Pig, Primates, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89115763 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-763 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-763 Supplier Abnova Corporation Supplier No. PAB12388
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of MAT2A.

Specifications

Antigen MAT2A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of MAT2A.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (30% glycerol, 0.09% sodium azide)
Gene MAT2A
Gene Alias MATA2/MATII/SAMS2
Gene Symbols MAT2A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1-100 of human MAT2A.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4144
Target Species Bovine, Chicken, Human, Mouse, Pig, Primate, Rat
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.