Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-MATE1, Polyclonal,

Catalog No. ABC522MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC522MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC522MI Supplier MilliporeSigma™ Supplier No. ABC522
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects MATE1 in Human samples, and it is validated for Western Blotting

MATE1 is part of our innate metabolism and functions as multi-substrate small molecule transporter. MATE1 is widely expressed in many tissues and is found especially found in luminal membranes of urinary tubules, bile caniculi and brush border membranes. MATE1 is the cellular transporter for tetraethylammonium (TEA), 1-methyl-4-phenylpyridinium (MPP), cimetidine, N-methylnicotinamide (NMN), metformin, creatinine, guanidine, procainamide, topotecan, estrone sulfate, cephalosporin, cephalexin and cephradin, and MATE1 often mediates the final excretion step for toxic organic cations. In disease treatments, it also is the transporter for the uptake of various anti-cancer drugs like acyclovir and ganciclovir but also toxins like Paraquat (a potent herbicide).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MATE1
Applications Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q96FL8
Gene Symbols SLC47A1; MATE1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the C-terminus of human MATE1.
Purification Method Affinity Purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_060712
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.