Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBD2 Rabbit anti-Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89116906 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-906 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-906 Supplier Abnova Corporation Supplier No. PAB14065
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Mbd2.

Specifications

Antigen MBD2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Mbd2.
Dilution ELISA (1:1000-1:4000) Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In serum (0.05% sodium azide)
Gene Mbd2
Gene Alias MBD2a
Gene Symbols Mbd2
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to internal region of mouse Mbd2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17191
Target Species Mouse
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.