Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBP (Maltose Binding Protein) Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89136909 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-909 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-909 Supplier Abnova Corporation Supplier No. PAB0893
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against MBP (Maltose Binding Protein).

Specifications

Antigen MBP (Maltose Binding Protein)
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against MBP (Maltose Binding Protein).
Dilution Western Blot (1:400) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% gelatin, 0.09% sodium azide)
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length MBP (Maltose Binding Protein).
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody detects fusion proteins containing MBP.
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.