Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mdm2 (phospho S185), Rabbit anti-Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89114797 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-797 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-797 Supplier Abnova Corporation Supplier No. PAB10334
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of Mdm2.

Specifications

Antigen Mdm2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of Mdm2.
Dilution ELISA (1:3000-1:12000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene Mdm2
Gene Alias 1700007J15Rik/AA415488/Mdm-2
Gene Symbols Mdm2
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S185 of mouse Mdm2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17246
Test Specificity Reactivity occurs against Mouse MDM2 pS185 protein and This antibody is specific to the phosphorylated form of the protein. Reactivity with non-phosphorylated mouse MDM2 is minimal by ELISA and western blot.
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.