Learn More
MED26 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MED26.
Supplier: Abnova Corporation PAB14124
Description
The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors. [provided by RefSeq
Specifications
MED26 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of MED26. | |
In PBS (0.05% sodium azide, 0.05% proclin 300) | |
CRSP7/CRSP70 | |
Rabbit | |
50 μg | |
Primary | |
Human | |
Liquid |
Western Blot | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
MED26 | |
MED26 | |
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human MED26. | |
RUO | |
9441 | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only