Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Rabbit anti-Menin/MEN1, Polyclonal,
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ ABC514
Description
Specifically detects Menin/MEN1 in Mouse samples, and it is validated for Western Blotting
Menin (UniProt O00255) is a nuclear protein encoded by the MEN1 gene (also known as MEAI, SCG2) (Gene ID 4221) in human. MEN1 is a tumor suppressor gene frequently mutated in the familial multiple endocrine neoplasia type I (MEN-1) cancer syndrome characterized by tumors in multiple endocrine organs including the parathyroid, pancreatic islets, and the pituitary. Menin mediates cell proliferation and differentiation actions of transforming growth factor-beta (TGF-beta) ligand family members. Menin is reported to associate with histone methyltransferases (HMT) MLL1 (KMT2A) and MLL4 (KMT2B) to form menin-HMT complexes in both human and murine model systems. In addition, menin is also shown to be involved in DNA repair with FANCD2.
Specifications
Menin/MEN1 | |
Polyclonal | |
O00255 | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
Mouse | |
Serum |
Western Blot | |
Rabbit polyclonal serum with 0.05% sodium azide. | |
MEN1; SCG2 | |
KLH-conjugated linear peptide corresponding to the C-terminus of human Menin/MEN1. | |
100 μL | |
Apoptosis, Cancer | |
NP_000235 | |
Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction