Learn More
MET Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MET.
Supplier: Abnova Corporation PAB12251
Description
The proto-oncogene MET product is the hepatocyte growth factor receptor and encodes tyrosine-kinase activity. The primary single chain precursor protein is post-translationally cleaved to produce the alpha and beta subunits, which are disulfide linked to form the mature receptor. Various mutations in the MET gene are associated with papillary renal carcinoma. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: VDYRASpecifications
MET | |
Polyclonal | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
MET | |
MET | |
A synthetic peptide (conjugated with KLH) corresponding to residues surrounding Y1003 of human MET. | |
100 μL | |
Primary | |
Met (Ab-1003) Antibody detects endogenous levels of total Met protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed) | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of MET. | |
In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
AUTS9/HGFR/RCCP2/c-Met | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
4233 | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only