Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylated Lysine, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89136962 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-962 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-962 Supplier Abnova Corporation Supplier No. PAB10350
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against Methylated Lysine.

Specifications

Antigen Methylated Lysine
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Methylated Lysine.
Dilution ELISA (1:5000) Western Blot (1:1000-1:5000) Immunoprecipitation (10 ug/mg sample) The optimal working dilution should be determined by the end user.
Formulation In 100mM Tris-Cl, 500mM NaCl, pH 8.0 (0.01% sodium azide)
Host Species Rabbit
Immunogen Methylated Lysine conjugated with KLH.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reactivity is specific to methylated lysine.
Target Species Mammalia
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.